missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoro-6-nitrophenol, 97%, Thermo Scientific™

Catalog Number 11466116
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
82419-26-9
C6H3F2NO3
175.091
MFCD00042255
KEGOHDCHURMFKX-UHFFFAOYSA-N
2,3-difluoro-6-nitrophenol, phenol, 2,3-difluoro-6-nitro, 2,3-difluoro-6-nitro-phenol, 6-nitro-2,3-difluorophenol, 2-hydroxy-3,4-difluoronitrobenzene, pubchem2873, acmc-209pot, 2,3 difluoro 6-nitrophenol, 2-nitro-5,6-difluorophenol, ksc495k9h
2733745
C1=CC(=C(C(=C1[N+](=O)[O-])O)F)F
This item is not returnable. View return policy
82419-26-9
C6H3F2NO3
175.091
MFCD00042255
KEGOHDCHURMFKX-UHFFFAOYSA-N
2,3-difluoro-6-nitrophenol, phenol, 2,3-difluoro-6-nitro, 2,3-difluoro-6-nitro-phenol, 6-nitro-2,3-difluorophenol, 2-hydroxy-3,4-difluoronitrobenzene, pubchem2873, acmc-209pot, 2,3 difluoro 6-nitrophenol, 2-nitro-5,6-difluorophenol, ksc495k9h
2733745
C1=CC(=C(C(=C1[N+](=O)[O-])O)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 60°C to 62°C
Quantity 1 g
Beilstein 4410368
Formula Weight 175.09
Percent Purity 97%
Chemical Name or Material 2,3-Difluoro-6-nitrophenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'