missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoro-4-methoxyphenol, 97%, Thermo Scientific™

Product Code. 11471160
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471160 1 g 1g
11481160 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11471160 Supplier Thermo Scientific Alfa Aesar Supplier No. H26305.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Analytical application of poly{methyl[3-(2-hydroxy-3,4-difluoro)phenyl]propyl siloxane} as a QCM coating for DMMP detection.

Solubility
Slightly Soluble (1.8 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261763-29-5
Molecular Formula C7H6F2O2
Molecular Weight (g/mol) 160.12
MDL Number MFCD01631505
InChI Key CKKAZWYLHGNVDZ-UHFFFAOYSA-N
Synonym phenol, 2,3-difluoro-4-methoxy, 1-hydroxy-4-methoxy-2,3-difluorobenzene, acmc-20aogz, 2,3-difluoro-4-hydroxyanisole, 2,3-difluoro-4-methoxy-phenol, 2,3-difluoro-4-methoxyphenol, phenol, 2,3-difluoro-4-methoxy-9ci
PubChem CID 2774120
IUPAC Name 2,3-difluoro-4-methoxyphenol
SMILES COC1=C(C(=C(C=C1)O)F)F

Specifications

Melting Point 103°C to 106°C
Quantity 1 g
Solubility Information Slightly Soluble (1.8g/L) (25°C).
Formula Weight 160.12
Percent Purity 97%
Chemical Name or Material 2,3-Difluoro-4-methoxyphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.