missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Difluoro-4-ethoxybenzeneboronic acid, 98%, Thermo Scientific™
£20.10 - £89.51
Chemical Identifiers
CAS | 212386-71-5 |
---|---|
Molecular Formula | C8H9BF2O3 |
Molecular Weight (g/mol) | 201.96 |
MDL Number | MFCD09258743 |
InChI Key | OBKSFBWOZSQGGC-UHFFFAOYSA-N |
Synonym | 2,3-difluoro-4-ethoxyphenylboronic acid, 4-ethoxy-2,3-difluorophenyl boronic acid, 2,3-difluoro-4-ethoxybenzeneboronic acid, boronic acid, 4-ethoxy-2,3-difluorophenyl, pubchem9562, 2,3-difluoro-4-ethoxyphenylboronicacid, acmc-209fik, ksc201g9b, 4-ethoxy-2,3-difluorobenzeneboronic acid |
PubChem CID | 15135235 |
IUPAC Name | (4-ethoxy-2,3-difluorophenyl)boronic acid |
SMILES | CCOC1=C(F)C(F)=C(C=C1)B(O)O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
212386-71-5 | |
201.96 | |
OBKSFBWOZSQGGC-UHFFFAOYSA-N | |
15135235 | |
CCOC1=C(F)C(F)=C(C=C1)B(O)O |
C8H9BF2O3 | |
MFCD09258743 | |
2,3-difluoro-4-ethoxyphenylboronic acid, 4-ethoxy-2,3-difluorophenyl boronic acid, 2,3-difluoro-4-ethoxybenzeneboronic acid, boronic acid, 4-ethoxy-2,3-difluorophenyl, pubchem9562, 2,3-difluoro-4-ethoxyphenylboronicacid, acmc-209fik, ksc201g9b, 4-ethoxy-2,3-difluorobenzeneboronic acid | |
(4-ethoxy-2,3-difluorophenyl)boronic acid |
Specifications
212386-71-5 | |
C8H9BF2O3 | |
19662857 | |
OBKSFBWOZSQGGC-UHFFFAOYSA-N | |
(4-ethoxy-2,3-difluorophenyl)boronic acid | |
15135235 | |
98% | |
2,3-Difluoro-4-ethoxybenzeneboronic acid |
151°C to 154°C | |
MFCD09258743 | |
2,3-difluoro-4-ethoxyphenylboronic acid, 4-ethoxy-2,3-difluorophenyl boronic acid, 2,3-difluoro-4-ethoxybenzeneboronic acid, boronic acid, 4-ethoxy-2,3-difluorophenyl, pubchem9562, 2,3-difluoro-4-ethoxyphenylboronicacid, acmc-209fik, ksc201g9b, 4-ethoxy-2,3-difluorobenzeneboronic acid | |
CCOC1=C(F)C(F)=C(C=C1)B(O)O | |
201.96 | |
201.96 | |
Solid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures