missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diethyl-5-methylpyrazine-N4-oxide, TRC

Product Code. 30379088
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30379088 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30379088 Supplier Toronto Research Chemicals Supplier No. TRCD44453010MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1076200-01-5
Molecular Formula C9 H14 N2 O
Quantity 10 mg
InChI Formula InChI=1S/C9H14N2O/c1-4-8-9(5-2)11(12)7(3)6-10-8/h6H,4-5H2,1-3H3
SMILES CCc1ncc(C)[n+]([O-])c1CC
IUPAC Name 2,3-diethyl-6-methyl-1-oxidopyrazin-1-ium
Molecular Weight (g/mol) 166.22
Formula Weight 166.1106
Chemical Name or Material 2,3-Diethyl-5-methylpyrazine-N4-oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.