missing translation for 'onlineSavingsMsg'
Learn More

2',3'-Dideoxyadenosine, MedChemExpress

Product Code. 16440479
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16440479 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16440479 Supplier Medchem Express Supplier No. HYW013441
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 2',3'-Dideoxyadenosine is an inhibitor of HIV replication. Antiretroviral activity. Antiviral efficacy.

2',3'-Dideoxyadenosine is an inhibitor of HIV replication. Antiretroviral activity. Antiviral efficacy.

Specifications

Chemical Name or Material 2',3'-Dideoxyadenosine
Physical Form Solid
CAS 4097-22-7
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C10H13N5O2
For Use With (Application) COVID-19-anti-virus
Solubility Information DMSO : 100 mg/mL (425.10 mM; Need ultrasonic) ∣H2O : 100 mg/mL (425.10 mM; Need ultrasonic)
SMILES NC1=C(N=CN2[C@H]3CC[C@H](O3)CO)C2=NC=N1
Molecular Weight (g/mol) 235.24
Formula Weight 235.24
Percent Purity 99.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.