missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dicyanohydroquinone, 98%

Product Code. 10143581 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10143581 25 g Glass bottle 25g
10557021 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10143581 Supplier Thermo Scientific Acros Supplier No. 232050250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4733-50-0 | C8H4N2O2 | 160.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4733-50-0
MDL Number MFCD00001790
InChI Key MPAIWVOBMLSHQA-UHFFFAOYSA-N
Synonym 2,3-dicyanohydroquinone, 3,6-dihydroxyphthalonitrile, 2,3-dicyano-p-hydroquinone, 3,6-dihydroxyphthalodinitrile, 1,2-benzenedicarbonitrile, 3,6-dihydroxy, 2,3-dicyanobenzene-1,4-diol, phthalonitrile, 3,6-dihydroxy, dicyanohydroquinone, 2,3-dicyanohydroquinme, acmc-1asrw
PubChem CID 78467
IUPAC Name 3,6-dihydroxybenzene-1,2-dicarbonitrile
SMILES C1=CC(=C(C(=C1O)C#N)C#N)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >230.0°C
Color Brown to Khaki
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C8H4N2O2
Linear Formula (NC)2C6H2-1,4-(OH)2
Quantity 25 g
Molecular Weight (g/mol) 160.132
Formula Weight 160.13
Percent Purity 98%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 2, 3-Dicyanohydroquinone, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2,3-Dicyanohydroquinone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.