missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorotoluene, 98%

Product Code. 11464110
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464110 50 g 50g
11493514 25 g 25g
11403524 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464110 Supplier Thermo Scientific Alfa Aesar Supplier No. A11206.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 32768-54-0 | C7H6Cl2 | 161.025 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32768-54-0
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.025
MDL Number MFCD00000546
InChI Key GWLKCPXYBLCEKC-UHFFFAOYSA-N
Synonym 2,3-dichlorotoluene, benzene, 1,2-dichloro-3-methyl, toluene, 2,3-dichloro, 1,2-dichloro-3-methyl-benzene, acmc-209hvi, ksc490m7b, gwlkcpxyblcekc-uhfffaoysa, 2,3-dct, #, labotest-bb ltbb004413
PubChem CID 34702
IUPAC Name 1,2-dichloro-3-methylbenzene
SMILES CC1=C(C(=CC=C1)Cl)Cl

Specifications

Melting Point 4°C
Density 1.229
Boiling Point 207°C to 208°C
Flash Point 83°C (181°F)
Refractive Index 1.551
Quantity 50 g
UN Number UN3082
Beilstein 2412384
Formula Weight 161.03
Percent Purity 98%
Chemical Name or Material 2,3-Dichlorotoluene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.