missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorothiophene-5-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 11362617
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 11362617

Brand: Thermo Scientific Alfa Aesar L11780.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126714-85-0
Molecular Formula C4HCl3O2S2
Molecular Weight (g/mol) 251.52
MDL Number MFCD00052092
InChI Key IVTWLTRKVRJPNG-UHFFFAOYSA-N
Synonym 2,3-dichlorothiophene-5-sulfonyl chloride, 2,3-dichlorothiophene-5-sulphonyl chloride, 4,5-dichloro-2-thiophenesulfonyl chloride, 4,5-dichloro-thiophene-2-sulfonyl chloride, 2-thiophenesulfonyl chloride, 4,5-dichloro, pubchem5523, acmc-20aoh3, 4,5-dichloro 2-thienyl chlorosulfone, 2,3-dichlorothiophene-5-sulfonylchloride, 2-thiophenesulfonylchloride, 4,5-dichloro
PubChem CID 518622
IUPAC Name 4,5-dichlorothiophene-2-sulfonyl chloride
SMILES ClC1=C(Cl)C=C(S1)S(Cl)(=O)=O

Specifications

Melting Point 54°C to 57°C
Quantity 1 g
UN Number UN3261
Beilstein 161983
Sensitivity Moisture sensitive
Formula Weight 251.54
Percent Purity 97%
Chemical Name or Material 2,3-Dichlorothiophene-5-sulfonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.