missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorophenylhydrazine hydrochloride, 97%

Product Code. 11454290
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454290 25 g 25g
11444290 5 g 5g
11464290 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454290 Supplier Thermo Scientific Alfa Aesar Supplier No. A10717.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21938-47-6 | C6H6Cl2N2 | 177.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21938-47-6
Molecular Formula C6H6Cl2N2
Molecular Weight (g/mol) 177.03
MDL Number MFCD00052265
InChI Key HPIALSXRVQRGMK-UHFFFAOYSA-N
Synonym 2,3-dichlorophenylhydrazine hydrochloride, 2,3-dichlorophenyl hydrazine hydrochloride, hydrazine, 2,3-dichlorophenyl-, monohydrochloride, 1-2,3-dichlorophenyl hydrazine hydrochloride, pubchem7561, acmc-209fpn, ksc494k0t, 2,3-dichlorophenylhydrazine, chloride, 2,3-dichlorophenylhydrazine hcl
PubChem CID 12276572
IUPAC Name (2,3-dichlorophenyl)hydrazine;hydrochloride
SMILES NNC1=CC=CC(Cl)=C1Cl

Specifications

Melting Point ∼204°C (decomposition)
Quantity 25 g
Beilstein 3708910
Formula Weight 213.49
Percent Purity 97%
Chemical Name or Material 2,3-Dichlorophenylhydrazine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.