missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorophenyl isothiocyanate 97%, Thermo Scientific™

Product Code. 10086604
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 6590-97-2
Molecular Formula C7H3Cl2NS
Molecular Weight (g/mol) 204.07
MDL Number MFCD00041048
InChI Key RQZIODPVCCTBAQ-UHFFFAOYSA-N
PubChem CID 2796281
IUPAC Name 1,2-dichloro-3-isothiocyanatobenzene
SMILES ClC1=CC=CC(N=C=S)=C1Cl

Specifications

Density 1.4340g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym 2,3-dichlorophenyl isothiocyanate, 2,3-dichlorophenylisothiocyanate, benzene,1,2-dichloro-3-isothiocyanato, benzene, 1,2-dichloro-3-isothiocyanato, dichlorophenylisothiocyanate, dichlorophenyl isothiocyanate, acmc-1b4g1, 2,3-dichlorophenyl-isothiocyanate, 1,2-dichloro-3-isothiocyanato-benzene, 2,3-dichloro-1-isothiocyanato-benzene
Formula Weight 204.08
Percent Purity 97%
Chemical Name or Material 2, 3-Dichlorophenyl isothiocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'