missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorophenyl isothiocyanate 97%, Thermo Scientific™

Product Code. 10225314
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10225314 25 g 25g
10086604 5 g 5g
10313113 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10225314 Supplier Thermo Scientific Maybridge Supplier No. SB01755EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 6590-97-2
Molecular Formula C7H3Cl2NS
Molecular Weight (g/mol) 204.07
MDL Number MFCD00041048
InChI Key RQZIODPVCCTBAQ-UHFFFAOYSA-N
PubChem CID 2796281
IUPAC Name 1,2-dichloro-3-isothiocyanatobenzene
SMILES ClC1=CC=CC(N=C=S)=C1Cl

Specifications

Density 1.434g/mL
Assay Percent Range 97%
Quantity 25 g
Synonym 2,3-dichlorophenyl isothiocyanate, 2,3-dichlorophenylisothiocyanate, benzene,1,2-dichloro-3-isothiocyanato, benzene, 1,2-dichloro-3-isothiocyanato, dichlorophenylisothiocyanate, dichlorophenyl isothiocyanate, acmc-1b4g1, 2,3-dichlorophenyl-isothiocyanate, 1,2-dichloro-3-isothiocyanato-benzene, 2,3-dichloro-1-isothiocyanato-benzene
Formula Weight 204.08
Percent Purity 97%
Chemical Name or Material 2, 3-Dichlorophenyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.