missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorophenyl isocyanate, 97%

Product Code. 11323686
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11323686 1 g 1g
11333686 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11323686 Supplier Thermo Scientific Alfa Aesar Supplier No. L14780.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 41195-90-8 | C7H3Cl2NO | 188.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41195-90-8
Molecular Formula C7H3Cl2NO
Molecular Weight (g/mol) 188.01
MDL Number MFCD00002000
InChI Key FYWJWWMKCARWQG-UHFFFAOYSA-N
Synonym 2,3-dichlorophenyl isocyanate, 2,3-dichlorophenylisocyanate, benzene, 1,2-dichloro-3-isocyanato, 1,2-dichloro-3-isocyanato-benzene, 2,3-dichlorobenzenisocyanate, benzene, dichloroisocyanato, dichlorphenylisocyanate, dichlorophenyl isocyanate, acmc-1ap4f, 2,3dichlorophenylisocyanate
PubChem CID 33050
IUPAC Name 1,2-dichloro-3-isocyanatobenzene
SMILES ClC1=CC=CC(N=C=O)=C1Cl

Specifications

Density 1.424
Boiling Point 58°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5784
Quantity 1 g
UN Number UN2206
Beilstein 4669222
Sensitivity Moisture sensitive
Formula Weight 188.01
Percent Purity 97%
Chemical Name or Material 2,3-Dichlorophenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.