missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorophenol, TRC

Product Code. 30371117
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30371117 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30371117 Supplier Toronto Research Chemicals Supplier No. TRCD436185100G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3-Dichlorophenol
CAS 576-24-9
Molecular Formula C6 H4 Cl2 O
Quantity 100 g
Synonym Phenol, 2,3-dichloro- (8CI, 9CI, ACI), 2,3-Dichlorophenol (ACI), NSC 60646
InChI Formula InChI=1S/C6H4Cl2O/c7-4-2-1-3-5(9)6(4)8/h1-3,9H
SMILES Oc1cccc(Cl)c1Cl
IUPAC Name 2,3-dichlorophenol
Molecular Weight (g/mol) 163
Formula Weight 161.9639
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.