missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorodibenzo-p-dioxin, TRC

Product Code. 30386795
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386795 5 mg
30379840 10 mg
30376827 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386795 Supplier Toronto Research Chemicals Supplier No. TRCD4340855MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29446-15-9
Molecular Formula C12 H6 Cl2 O2
Molecular Weight (g/mol) 253.08
Synonym Dibenzo[b,e][1,4]dioxin, 2,3-dichloro-, Dibenzo-p-dioxin, 2,3-dichloro- (8CI), 2,3-Dichlorodibenzo[b,e][1,4]dioxin, 2,3-Dichlorodibenzo-p-dioxin, 2,3-Dichlorodibenzodioxin, PCDD 10
IUPAC Name 2,3-dichlorodibenzo-p-dioxin
SMILES Clc1cc2Oc3ccccc3Oc2cc1Cl

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C12H6Cl2O2/c13-7-5-11-12(6-8(7)14)16-10-4-2-1-3-9(10)15-11/h1-6H
Formula Weight 251.9745
Chemical Name or Material 2,3-Dichlorodibenzo-p-dioxin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.