missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number 11317466
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
2905-60-4
C7H3Cl3O
209.45
MFCD00035937
YBONBWJSFMTXLE-UHFFFAOYSA-N
benzoyl chloride, dichloro, dichlorobenzoyl chloride, benzoyl chloride, 2,3-dichloro, unii-i50f21ps1n, 2,3-dichlorobenzoylchloride, acmc-1cfin, 2,3-dichlorobenzoyl-chloride, ksc202s8t, 2, 3-dichlorobenzoyl chloride, 2,3-dcoc
17944
2,3-dichlorobenzoyl chloride
ClC(=O)C1=CC=CC(Cl)=C1Cl
This item is not returnable. View return policy
2905-60-4
C7H3Cl3O
209.45
MFCD00035937
YBONBWJSFMTXLE-UHFFFAOYSA-N
benzoyl chloride, dichloro, dichlorobenzoyl chloride, benzoyl chloride, 2,3-dichloro, unii-i50f21ps1n, 2,3-dichlorobenzoylchloride, acmc-1cfin, 2,3-dichlorobenzoyl-chloride, ksc202s8t, 2, 3-dichlorobenzoyl chloride, 2,3-dcoc
17944
2,3-dichlorobenzoyl chloride
ClC(=O)C1=CC=CC(Cl)=C1Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 28°C to 32°C
Boiling Point 140°C (14 mmHg)
Flash Point 167°C (332°F)
Odor Pungent
Quantity 5 g
UN Number UN3261
Beilstein 2575973
Sensitivity Moisture sensitive
Formula Weight 209.46
Percent Purity 98%
Chemical Name or Material 2,3-Dichlorobenzoyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'