missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzonitrile, 98%

Product Code. 11472557
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472557 25 g 25g
11462557 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11472557 Supplier Thermo Scientific Alfa Aesar Supplier No. A14809.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6574-97-6 | C7H3Cl2N | 172.008 g/mol

2,3-Dichlorobenzonitrile, is used as a reactant involved in nucleophilic aromatic fluorination, reactions with magnesium amides for synthesis of carboxamides, Suzuki-Miyaura coupling reactions. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dichlorobenzonitrile, is used as a reactant involved in nucleophilic aromatic fluorination, reactions with magnesium amides for synthesis of carboxamides, Suzuki-Miyaura coupling reactions. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

Solubility
It is miscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6574-97-6
Molecular Formula C7H3Cl2N
Molecular Weight (g/mol) 172.008
MDL Number MFCD00016372
InChI Key OHDYZVVLNPXKDX-UHFFFAOYSA-N
Synonym benzonitrile, dichloro, 2,3-dichlorobenzenecarbonitrile, benzonitrile, 2,3-dichloro, dichlorobenzonitrile, pubchem12516, 2,3-dichloro benzonitrile, 2,3-dichlorophenyl cyanide, 2,3-di-chloro-benzonitrile, acmc-1b38y, ksc357s4b
PubChem CID 736567
IUPAC Name 2,3-dichlorobenzonitrile
SMILES C1=CC(=C(C(=C1)Cl)Cl)C#N

Specifications

Melting Point 60°C to 65°C
Boiling Point 114°C (6 mmHg)
Quantity 25 g
UN Number UN3439
Beilstein 2438753
Solubility Information It is miscible with water.
Formula Weight 172.01
Percent Purity 98%
Chemical Name or Material 2,3-Dichlorobenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.