missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzenesulfonamide (>90%), TRC

Product Code. 30377494
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30377494 50 mg
30376023 500 mg
30382367 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30377494 Supplier Toronto Research Chemicals Supplier No. TRCD43534350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82967-94-0
Molecular Formula C6H5NO2SCl2
Molecular Weight (g/mol) 226.08
Synonym 2,3-Dichlorobenzenesulfonamide, 2,3-Dichlorobenzene-1-sulfonamide, M and B 9811
IUPAC Name 2,3-dichlorobenzenesulfonamide
SMILES NS(=O)(=O)c1cccc(Cl)c1Cl

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C6H5Cl2NO2S/c7-4-2-1-3-5(6(4)8)12(9,10)11/h1-3H,(H2,9,10,11)
Formula Weight 224.9418
Chemical Name or Material 2,3-Dichlorobenzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.