missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzeneboronic acid, 98%

Product Code. 11499817
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499817 25 g 25g
11479817 1 g 1g
11489817 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499817 Supplier Thermo Scientific Alfa Aesar Supplier No. B22781.14
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 151169-74-3 | C6H5BCl2O2 | 190.81 g/mol

2,3-Dichlorobenzeneboronic acid is used as a reactant for Suzuki-Miyaura coupling reactions. It is also used in preparation of biologically and pharmacologically active molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dichlorobenzeneboronic acid is used as a reactant for Suzuki-Miyaura coupling reactions. It is also used in preparation of biologically and pharmacologically active molecules.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151169-74-3
Molecular Formula C6H5BCl2O2
Molecular Weight (g/mol) 190.81
MDL Number MFCD01075703
InChI Key TYIKXPOMOYDGCS-UHFFFAOYSA-N
Synonym 2,3-dichlorophenyl boronic acid, 2,3-dichlorobenzeneboronic acid, 2,3-dichlorophenyl boranediol, boronic acid, 2,3-dichlorophenyl, contains varying amounts of anhydride, pubchem1810, dichlorophenylboronic acid, acmc-1c0ur, dichlorobenzene boronic acid
PubChem CID 2734661
IUPAC Name (2,3-dichlorophenyl)boronic acid
SMILES OB(O)C1=C(Cl)C(Cl)=CC=C1

Specifications

Melting Point 233°C to 242°C
Quantity 25 g
Solubility Information Soluble in methanol.
Formula Weight 190.82
Percent Purity 98%
Chemical Name or Material 2,3-Dichlorobenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.