missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzaldehyde, TRC

Product Code. 30382732
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30382732 50 g 50g
30384493 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30382732 Supplier Toronto Research Chemicals Supplier No. TRCD43191550G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3-Dichlorobenzaldehyde
CAS 6334-18-5
Molecular Formula C7 H4 Cl2 O
Quantity 50 g
Synonym Benzaldehyde, 2,3-dichloro-, 2,3-Dichlorobenzaldehyde, NSC 28450
InChI Formula InChI=1S/C7H4Cl2O/c8-6-3-1-2-5(4-10)7(6)9/h1-4H
SMILES Clc1cccc(C=O)c1Cl
IUPAC Name 2,3-dichlorobenzaldehyde
Molecular Weight (g/mol) 175.01
Formula Weight 173.9639
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.