missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloroanisole, 96%, ACROS Organics™

Product Code. 10363013 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10363013 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10363013 Supplier Thermo Scientific Acros Supplier No. 168900250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1984-59-4
CAS Min % 95.0
CAS Max % 100.0
Melting Point 30°C to 35°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Molecular Formula C7H6Cl2O
Linear Formula Cl2C6H3OCH3
MDL Number MFCD00000545
Quantity 25 g
Beilstein 06, I, 102
Synonym 2,3-dichloroanisole, benzene, 1,2-dichloro-3-methoxy, 2,3-dichloromethoxybenzene, 2,3-dichloranisol, dichloroanisole 2,3-, 1,2-dichloro-3-methoxy-benzene, dichloroanisol, 2,3-dichloro-anisol, pubchem3680, 2,3-dichloro anisole
InChI Key HFEASCCDHUVYKU-UHFFFAOYSA-N
SMILES COC1=CC=CC(Cl)=C1Cl
IUPAC Name 1,2-dichloro-3-methoxybenzene
Molecular Weight (g/mol) 177.02
PubChem CID 16126
Formula Weight 177.03
Percent Purity ≥95%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 2, 3-Dichloroanisole, 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.