missing translation for 'onlineSavingsMsg'
Learn More

2',3'-Dichloroacetophenone, 98%

Product Code. 11430543
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430543 5 g 5g
11420543 1 g 1g
11440543 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430543 Supplier Thermo Scientific Alfa Aesar Supplier No. A12667.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56041-57-7 | C8H6Cl2O | 189.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56041-57-7
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.04
MDL Number MFCD00052988
InChI Key KMABBMYSEVZARZ-UHFFFAOYSA-N
Synonym 1-2,3-dichlorophenyl ethanone, 2',3'-dichloroacetophenone, 2,3-dichloroacetophenone, 1-2,3-dichlorophenyl ethan-1-one, ethanone, 1-2,3-dichlorophenyl, 1-acetyl-2,3-dichlorobenzene, pubchem3368, 1-2,3-dichlorophenyl-1-ethanone, 1-2,3-bis chloranyl phenyl ethanone
PubChem CID 91986
IUPAC Name 1-(2,3-dichlorophenyl)ethanone
SMILES CC(=O)C1=CC=CC(Cl)=C1Cl

Specifications

Density 1.293
Boiling Point 106°C to 108°C (2 mmHg)
Refractive Index 1.56
Quantity 5 g
Beilstein 2438815
Formula Weight 189.04
Percent Purity 98%
Chemical Name or Material 2',3'-Dichloroacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.