missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-N'-hydroxybenzenecarboximidamide, 97%, Thermo Scientific™

Product Code. 10364062
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10364062 1 g 1g
10497822 250 mg 250mg
10515954 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10364062 Supplier Thermo Scientific Maybridge Supplier No. SPB01106DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 261761-55-1
Molecular Formula C7H6Cl2N2O
Molecular Weight (g/mol) 205.038
MDL Number MFCD01567248
InChI Key OPOPCNCEDLBYPV-UHFFFAOYSA-N
Synonym 2,3-dichlorobenzamidoxime, hcmhpiwpbbblbbbftocadcpba`, 2,3-dichlorophenyl hydroxyimino methylamine, z-2,3-dichloro-n'-hydroxybenzene-1-carboximidamide
PubChem CID 9580371
IUPAC Name 2,3-dichloro-N'-hydroxybenzenecarboximidamide
SMILES C1=CC(=C(C(=C1)Cl)Cl)C(=NO)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula Cl2C6H3C(=NOH)NH2
Quantity 1 g
Formula Weight 205.04
Percent Purity 97%
Chemical Name or Material 2, 3-Dichloro-N'-hydroxybenzenecarboximidamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.