missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-6-fluorophenylacetic acid, 97%, Thermo Scientific™

Catalog Number 15403457
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
886497-57-0
C8H5Cl2FO2
223.024
MFCD04115875
SQADAIPGCGGTQM-UHFFFAOYSA-N
2,3-dichloro-6-fluorophenylacetic acid, 2-2,3-dichloro-6-fluorophenyl acetic acid, 2,3-dichloro-6-fluorophenyl acetic acid, benzeneacetic acid,2,3-dichloro-6-fluoro
3511391
2-(2,3-dichloro-6-fluorophenyl)acetic acid
C1=CC(=C(C(=C1F)CC(=O)O)Cl)Cl
This item is not returnable. View return policy
886497-57-0
C8H5Cl2FO2
223.024
MFCD04115875
SQADAIPGCGGTQM-UHFFFAOYSA-N
2,3-dichloro-6-fluorophenylacetic acid, 2-2,3-dichloro-6-fluorophenyl acetic acid, 2,3-dichloro-6-fluorophenyl acetic acid, benzeneacetic acid,2,3-dichloro-6-fluoro
3511391
2-(2,3-dichloro-6-fluorophenyl)acetic acid
C1=CC(=C(C(=C1F)CC(=O)O)Cl)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 131°C to 135°C
Quantity 250 mg
Formula Weight 223.03
Percent Purity 97%
Chemical Name or Material 2,3-Dichloro-6-fluorophenylacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'