missing translation for 'onlineSavingsMsg'
Learn More

(2,3-Dichloro-4-oxyphenyl)-2-thienylmethanone, TRC

Product Code. 30370968
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30370968 500 mg -
30383593 250 mg -
30380176 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30370968 Supplier Toronto Research Chemicals Supplier No. TRCD435675500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40180-03-8
Molecular Formula C11 H6 Cl2 O2 S
Molecular Weight (g/mol) 273.14
IUPAC Name (2,3-dichloro-4-hydroxyphenyl)-thiophen-2-ylmethanone
SMILES Oc1ccc(C(=O)c2cccs2)c(Cl)c1Cl

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C11H6Cl2O2S/c12-9-6(3-4-7(14)10(9)13)11(15)8-2-1-5-16-8/h1-5,14H
Formula Weight 271.9466
Chemical Name or Material (2,3-Dichloro-4-oxyphenyl)-2-thienylmethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.