missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-1-propene, 98%

Product Code. 11312796
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312796 10 g 10g
11322796 50 g 50g
15488889 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11312796 Supplier Thermo Scientific Alfa Aesar Supplier No. L08943.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 78-88-6 | C3H4Cl2 | 110.965 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water (2g/L).

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-88-6
Molecular Formula C3H4Cl2
Molecular Weight (g/mol) 110.965
MDL Number MFCD00000943
InChI Key FALCMQXTWHPRIH-UHFFFAOYSA-N
Synonym 2,3-dichloropropene, 2,3-dichloro-1-propene, 2-chloroallyl chloride, 2,3-dichloropropylene, 1-propene, 2,3-dichloro, propene, 2,3-dichloro, 1,2-dichloro-2-propene, propylene, 2,3-dichloro, ccris 956, unii-370tnk5i41
PubChem CID 6565
IUPAC Name 2,3-dichloroprop-1-ene
SMILES C=C(CCl)Cl

Specifications

Melting Point -83°C
Boiling Point 93°C to 96°C
Flash Point 10°C (50°F)
Odor Pungent
Linear Formula ClCH2CCl=CH2
Quantity 10 g
UN Number UN2047
Beilstein 1361491
Formula Weight 110.97
Percent Purity 98%
Chemical Name or Material 2,3-Dichloro-1-propene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.