missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-1-propanol, TRC

Product Code. 30385060
Change view
Click to view available options
Quantity:
100 mg
25 g
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385060 25 g -
30375515 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30385060 Supplier Toronto Research Chemicals Supplier No. TRCD43577525G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-23-9
Molecular Formula C3 H6 Cl2 O
Molecular Weight (g/mol) 128.99
Synonym 2,3-Dichloro-1-propanol, (RS)-2,3-Dichloro-1-propanol, (±)-2,3-Dichloro-1-propanol, 1,2-Dichlorohydrin, 2,3-Dichlorohydrin, 2,3-Dichloropropyl alcohol, Glycerol 1,2-dichlorohydrin, Glycerol α,β-dichlorohydrin, α,β-Dichlorohydrin, β,γ-Dichlorohydrin, β-Dichlorohydrin
IUPAC Name 2,3-dichloropropan-1-ol
SMILES OCC(Cl)CCl

Specifications

Quantity 25 g
InChI Formula InChI=1S/C3H6Cl2O/c4-1-3(5)2-6/h3,6H,1-2H2
Formula Weight 127.9796
Chemical Name or Material 2,3-Dichloro-1-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.