missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dichloro-1,4-naphthoquinone, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Dichlone |
| CAS | 117-80-6 |
| Molecular Formula | C10 H4 Cl2 O2 |
| Quantity | 25 g |
| Synonym | 1,4-Naphthoquinone, 2,3-dichloro- (8CI), 2,3-Dichloro-1,4-naphthalenedione, 2,3-Dichloro-1,4-dihydro-1,4-dioxonaphthalene, 2,3-Dichloro-1,4-naphthoquinone, 2,3-Dichloro-p-naphthoquinone, Algistat, Algistat (quinone), Compound 604, Dichlone, Diclone, NSC 537, Phygon, Phygon Seed Protectant, Phygon XL, Sanquinon, USR 604 |
| InChI Formula | InChI=1S/C10H4Cl2O2/c11-7-8(12)10(14)6-4-2-1-3-5(6)9(7)13/h1-4H |
| SMILES | ClC1=C(Cl)C(=O)c2ccccc2C1=O |
| IUPAC Name | 2,3-dichloronaphthalene-1,4-dione |
| Molecular Weight (g/mol) | 227.04 |
| Formula Weight | 225.9588 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction