missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromothiophene, 97%, Thermo Scientific™

Product Code. 10667231 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10667231 5 g Glass bottle 5g
10626851 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10667231 Supplier Thermo Scientific Acros Supplier No. 258760050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3140-93-0
MDL Number MFCD00005418
InChI Key ATRJNSFQBYKFSM-UHFFFAOYSA-N
Synonym thiophene, 2,3-dibromo, 2,3-dibromo thiophene, dibromothiophene, 2,3-dibromo-thiophene, 2,3-dibromthiophen, 2 3-dibromothiophene, pubchem5511, thiophene,3-dibromo, acmc-1ckid, ksc491i5b
PubChem CID 76590
IUPAC Name 2,3-dibromothiophene
SMILES C1=CSC(=C1Br)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Brown or Khaki-Orange to Yellow
Density 2.2000g/mL
Boiling Point 130°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C4H2Br2S
Refractive Index 1.6300 to 1.632
Quantity 5 g
Specific Gravity 2.2
Molecular Weight (g/mol) 241.92
Formula Weight 241.92
Percent Purity 97%
Physical Form Chunks and/or Powder
Chemical Name or Material 2, 3-Dibromothiophene, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.