missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromopyridine, 97%

Product Code. 11388896
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11388896 1 g 1g
11398896 5 g 5g
11308906 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11388896 Supplier Thermo Scientific Alfa Aesar Supplier No. L19527.03
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13534-89-9 | C5H3Br2N | 236.894 g/mol

It is employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Moisture sensitive. Store in dry condition. Keep oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13534-89-9
Molecular Formula C5H3Br2N
Molecular Weight (g/mol) 236.894
MDL Number MFCD00234014
InChI Key SLMHHOVQRSSRCV-UHFFFAOYSA-N
Synonym pyridine, 2,3-dibromo, dibromopyridine, 2,3-dibromo pyridine, pubchem5402, acmc-209c0b, 2,3-dibromopyridine, ksc492s9r, tpc-py084, slmhhovqrssrcv-uhfffaoysa, abbypharma ap-12-5128
PubChem CID 817102
IUPAC Name 2,3-dibromopyridine
SMILES C1=CC(=C(N=C1)Br)Br

Specifications

Melting Point 58°C to 60°C
Boiling Point 249°C to 250°C
Quantity 1 g
Beilstein 109828
Solubility Information Insoluble in water.
Formula Weight 236.9
Percent Purity 97%
Chemical Name or Material 2,3-Dibromopyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.