missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromobutane, (+/-) + meso, 98+%

Product Code. 11479552
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479552 5 g 5g
11489552 25 g 25g
11499552 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479552 Supplier Thermo Scientific Alfa Aesar Supplier No. A10219.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5408-86-6 | C4H8Br2 | 215.916 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5408-86-6
Molecular Formula C4H8Br2
Molecular Weight (g/mol) 215.916
MDL Number MFCD00000146
InChI Key BXXWFOGWXLJPPA-UHFFFAOYSA-N
Synonym butane, 2,3-dibromo, dl-2,3-dibromobutane, +/--2,3-dibromobutane, ss-butylene bromide, .+-.-2,3-dibromobutane, dibromo butane, butane,3-dibromo, beta-butylene bromide, r*,r*-+--2,3-dibromobutane, pubchem12534
PubChem CID 21508
IUPAC Name 2,3-dibromobutane
SMILES CC(C(C)Br)Br

Specifications

Density 1.76
Boiling Point 166°C to 167°C
Flash Point 123°C (253°F)
Refractive Index 1.5126
Quantity 5 g
Beilstein 1718916
Formula Weight 215.93
Percent Purity ≥98%
Chemical Name or Material 2,3-Dibromobutane, (±) + meso

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.