missing translation for 'onlineSavingsMsg'
Learn More

2,3'-Dibromoacetophenone, 97%

Product Code. 11462228
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462228 25 g 25g
11442228 1 g 1g
11452228 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11462228 Supplier Thermo Scientific Alfa Aesar Supplier No. A15210.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18523-22-3 | C8H6Br2O | 277.943 g/mol

It is employed as an intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an intermediate for pharmaceutical and organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18523-22-3
Molecular Formula C8H6Br2O
Molecular Weight (g/mol) 277.943
MDL Number MFCD00082698
InChI Key MZBXSQBQPJWECM-UHFFFAOYSA-N
Synonym 3-bromophenacyl bromide, 2,3'-dibromoacetophenone, 2-bromo-1-3-bromophenyl ethanone, ethanone, 2-bromo-1-3-bromophenyl, 2-bromo-1-3-bromophenyl ethan-1-one, 3-bromophenacylbromide, 2-bromo-1-3-bromo-phenyl-ethanone, 2-bromo-3'-bromoacetophenone, 2-bromo-1-3-bromophenyl-1-ethanone
PubChem CID 519585
IUPAC Name 2-bromo-1-(3-bromophenyl)ethanone
SMILES C1=CC(=CC(=C1)Br)C(=O)CBr

Specifications

Melting Point 49°C to 52°C
Odor Pungent
Quantity 25 g
UN Number UN3261
Beilstein 775786
Solubility Information Insoluble in water.
Formula Weight 277.95
Percent Purity 97%
Chemical Name or Material 2,3'-Dibromoacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.