missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromo-3-(2-bromophenyl)propionic Acid, TRC

Product Code. 30381670
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381670 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381670 Supplier Toronto Research Chemicals Supplier No. TRCD4251152G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3-Dibromo-3-(2-bromophenyl)propionic Acid
CAS 246876-04-0
Molecular Formula C9 H7 Br3 O2
Quantity 2 g
InChI Formula InChI=1S/C9H7Br3O2/c10-6-4-2-1-3-5(6)7(11)8(12)9(13)14/h1-4,7-8H,(H,13,14)
SMILES OC(=O)C(Br)C(Br)c1ccccc1Br
IUPAC Name 2,3-dibromo-3-(2-bromophenyl)propanoic acid
Molecular Weight (g/mol) 386.86
Formula Weight 383.7996
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.