missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diaminopyridine, 98%

Product Code. 10112971 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10112971 5 g Glass bottle 5g
10216060 10 g Glass bottle 10g
10336651 25 g Glass bottle 25g
10309333 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10112971 Supplier Thermo Scientific Acros Supplier No. 155020050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 452-58-4 | C5H7N3 | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-58-4
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006319
InChI Key ZZYXNRREDYWPLN-UHFFFAOYSA-N
Synonym 2,3-diaminopyridine, 2,3-pyridinediamine, pyridine, 2,3-diamino, pyridine-2,3-diyldiamine, unii-cbx394737h, diaminopyridine, 2,3-diamino-pyridine, pyridinediamine, pyridine,3-diamino, pubchem1296
PubChem CID 9956
IUPAC Name pyridine-2,3-diamine
SMILES NC1=CC=CN=C1N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 114°C to 116°C
Flash Point 205°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (C5H3N)(NH2)2
Quantity 5 g
Beilstein 22,II,394
Solubility Information Solubility in water: 100g/L (20°C)
Formula Weight 109.13
Percent Purity 98%
Chemical Name or Material 2, 3-Diaminopyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2,3-Diaminopyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.