missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Diaminopropionic acid hydrochloride, MedChemExpress
Description
2,3-Diaminopropionic acid hydrochloride is a competitive inhibitor of cystathionase (CTH).
Specifications
Specifications
| Chemical Name or Material | 2,3-Diaminopropionic acid hydrochloride |
| CAS | 1482-97-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C3H9ClN2O2 |
| Quantity | 100 mg |
| Solubility Information | H2O : 100 mg/mL (711.39 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (7.11 mM; ultrasonic and warming and heat to 80°C) |
| SMILES | N[C@@H](CN)C(O)=O.Cl |
| Molecular Weight (g/mol) | 140.57 |
| Formula Weight | 140.57 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?