missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diaminonaphthalene, 97%

Product Code. 11479873
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479873 1 g 1g
11489873 5 g 5g
11499873 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479873 Supplier Thermo Scientific Alfa Aesar Supplier No. A12993.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 771-97-1 | C10H10N2 | 158.204 g/mol

2,3 Diaminonaphthalene is a highly selective colorimetric and fluorometric reagent for selenium detection and also used for the fluorometric determination of nitrite. It is also useful for detection of ketones and alpha-keto acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3 Diaminonaphthalene is a highly selective colorimetric and fluorometric reagent for selenium detection and also used for the fluorometric determination of nitrite. It is also useful for detection of ketones and alpha-keto acids.

Solubility
Slightly soluble in water. Soluble in pyridine, dimethylsulfoxide, dimethyformamide, dil.hydrochloric acid, ethanol, ether and hot methanol.

Notes
Incompatible with oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771-97-1
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.204
MDL Number MFCD00004116
InChI Key XTBLDMQMUSHDEN-UHFFFAOYSA-N
Synonym 2,3-diaminonaphthalene, 2,3-naphthalenediamine, 2,3-naphthylenediamine, unii-2bnz6brs87, 2bnz6brs87, ccris 8399, zlchem 848, 2,3-diaminonapthalene, 2,3 diaminonapthalene, pubchem15532
PubChem CID 69872
IUPAC Name naphthalene-2,3-diamine
SMILES C1=CC=C2C=C(C(=CC2=C1)N)N

Specifications

Melting Point 193°C to 196°C
Density 1.096
Quantity 1 g
Beilstein 2206394
Solubility Information Slightly soluble in water; Soluble in pyridine,dimethylsulfoxide,dimethyformamide,dil;hydrochloric acid,ethanol,ether and hot methanol.
Formula Weight 158.2
Percent Purity 97%
Chemical Name or Material 2,3-Diaminonaphthalene
compliance-icons
Product Identifier
  • 2,3-Naphthalenediamine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2,3-Naphthalenediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.