Learn More
2,3-Diaminonaphthalene, 97%, Thermo Scientific Chemicals
£41.70 - £456.00
Chemical Identifiers
CAS | 771-97-1, 91-59-8 |
---|---|
Molecular Formula | C10H10N2 |
Molecular Weight (g/mol) | 158.2 |
InChI Key | XTBLDMQMUSHDEN-UHFFFAOYSA-N |
Synonym | 2,3-diaminonaphthalene, 2,3-naphthalenediamine, 2,3-naphthylenediamine, unii-2bnz6brs87, 2bnz6brs87, ccris 8399, zlchem 848, 2,3-diaminonapthalene, 2,3 diaminonapthalene, pubchem15532 |
PubChem CID | 69872 |
IUPAC Name | naphthalene-2,3-diamine |
SMILES | C1=CC=C2C=C(C(=CC2=C1)N)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10001870
|
Thermo Scientific Acros
159590010 |
1 g | Glass bottle |
£41.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10347910
|
Thermo Scientific Acros
159590050 |
5 g | Glass bottle |
£196.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11915271
|
Thermo Scientific Acros
159590250 |
25 g | Glass bottle |
£456.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
771-97-1, 91-59-8 | |
158.2 | |
2,3-diaminonaphthalene, 2,3-naphthalenediamine, 2,3-naphthylenediamine, unii-2bnz6brs87, 2bnz6brs87, ccris 8399, zlchem 848, 2,3-diaminonapthalene, 2,3 diaminonapthalene, pubchem15532 | |
naphthalene-2,3-diamine |
C10H10N2 | |
XTBLDMQMUSHDEN-UHFFFAOYSA-N | |
69872 | |
C1=CC=C2C=C(C(=CC2=C1)N)N |
Specifications
771-97-1, 91-59-8 | |
Brown to Brown-Green | |
>110°C | |
96% min. (HPLC) | |
C10H6(NH2)2 | |
1.0968 | |
Solubility in water: < 0.1% (20°C). | |
C1=CC=C2C=C(C(=CC2=C1)N)N | |
158.2 | |
158.2 | |
Crystalline Powder |
193.0°C to 199.0°C | |
1.0968g/mL | |
Authentic | |
C10H10N2 | |
13,207 | |
2,3-diaminonaphthalene, 2,3-naphthalenediamine, 2,3-naphthylenediamine, unii-2bnz6brs87, 2bnz6brs87, ccris 8399, zlchem 848, 2,3-diaminonapthalene, 2,3 diaminonapthalene, pubchem15532 | |
XTBLDMQMUSHDEN-UHFFFAOYSA-N | |
naphthalene-2,3-diamine | |
69872 | |
97% | |
2,3-Diaminonaphthalene |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 212-241-
RUO – Research Use Only