missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diaminobenzoic acid, 95%

Product Code. 15545976 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15545976 1 g 1g
15555976 5 g 5g
15565976 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15545976 Supplier Thermo Scientific Acros Supplier No. 457490010
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 603-81-6 | C7H8N2O2 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-81-6
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00137818
InChI Key KKTUQAYCCLMNOA-UHFFFAOYSA-N
Synonym diaminobenzoic acid, benzoic acid, diamino, 3-carboxy-1,2-diaminobenzene, pubchem3145, acmc-209mjn, 2,3-diaminobenzonic acid, 2,3-diamino benzoic acid, ksc203g4l, pharmabridge p-2952, 2,3-diaminobenzoic acid
PubChem CID 198069
IUPAC Name 2,3-diaminobenzoic acid
SMILES NC1=CC=CC(C(O)=O)=C1N

Specifications

Melting Point 232.0°C to 234.5°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 152.15
Percent Purity 95%
Chemical Name or Material 2,3-Diaminobenzoic acid
compliance-icons
Product Identifier
  • 2,3-Diaminobenzoic acid, 95%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.