missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ 2,3-DCPE hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
2,3-DCPE hydrochloride is a selectively induces apoptosis and downregulates Bcl-XL protein expression in various human cancer cells versus normal cells in vitro. IC50 values are 0.89 and 12.6 μM for LoVo human colon cancer cell line and normal human fibroblasts respectively. Induces p21 expression and S-phase arrest in cancer cells via ERK-mediated pathways.
Specifications
Specifications
| Purity | >99% |
| Quantity | 10 mg |
| Product Type | 2,3-DCPE hydrochloride |
| Molecular Weight (g/mol) | 300.61 |
| Inhibitors | 2,3-DCPE hydrochloride |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction