Learn More
Alfa Aesar™ 2-(3-Chlorophenyl)benzimidazole, 95%
Brand: Alfa Aesar™ H64245.03
Code : NEW
Additional Details : CAS Number : 22868-35-5 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
2-(3-Chlorophenyl)benzimidazole | |
22868-35-5 | |
228.68 | |
MFCD01438282 |
Specifications
2-(3-Chlorophenyl)benzimidazole | |
22868-35-5 | |
95% | |
MFCD01438282 |
1g | |
Solid | |
228.68 |