missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 2',3'-cGAMP sodium salt
GREENER_CHOICE

Product Code. 18798688
Change view
Click to view available options
Quantity:
359 μg
Unit Size:
359µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18798688 359 μg 359µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18798688 Supplier Tocris Bioscience™ Supplier No. 5945/359U

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Endogenous high affinity STING agonist

2',3'-cGAMP sodium salt is a endogenous high affinity STING agonist (Kd values are 3.79 and 120 nM for human and rat STING, respectively). Induces repolarization of M2 macrophages in vitro.

This product is sold in units of 359μg, equivalent to 0.5μmol.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2′,3′-Cyclic guanosine monophosphate-adenosine monophosphate sodium salt
CAS 2734858-36-5
Synonym endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membr
Quantity 359 μg
Target STING/TMEM173
Molecular Formula C20H24N10Na2O13P2
Formula Weight 718.37
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.