missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Butynyloxy)tetrahydropyran, 98%

Product Code. 15460148
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15460148 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15460148 Supplier Alfa Aesar™ Supplier No. H53385.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Applications
2-(3-Butynyloxy)tetrahydropyran is used as a reagent in the synthesis of Phomoidride D, a bridged, polycyclic, natural compound that has the ability to inhibit squalene synthase and can be used as an antihypercholesterolemic agent. It is also used as a reagent to prepare Rugulactone, a natural compound that is isolated from Cryptocarya rugulosa.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40365-61-5
Molecular Formula C9H14O2
Molecular Weight (g/mol) 154.209
MDL Number MFCD00012352
InChI Key ZQZSNKJFOFAJQX-UHFFFAOYSA-N
Synonym 2-but-3-yn-1-yloxy tetrahydro-2h-pyran, 2-3-butynyloxy tetrahydro-2h-pyran, 2h-pyran, 2-3-butynyloxy tetrahydro, 2-3-butynyloxy tetrahydro-2 h-pyran, 1-tetrahydropyran-2-yloxy-3-butyne, 2-but-3-ynoxytetrahydropyran, tetrahydropyranyloxy-3-butyne, 2-but-3-yn-1-yloxy oxane, 2-3-butynyloxy tetrahydropyran, 4-tetrahydropyranyl oxy butyne
PubChem CID 142440
IUPAC Name 2-but-3-ynoxyoxane
SMILES C#CCCOC1CCCCO1

Specifications

Boiling Point 92°C to 95°C (18 mmHg)
Flash Point 73°C (163°F)
Quantity 25 g
Formula Weight 154.21
Percent Purity 98%
Chemical Name or Material 2-(3-Butynyloxy)tetrahydropyran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.