missing translation for 'onlineSavingsMsg'
Learn More

2,3-Butanedione, TRC

Product Code. 30134450
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Unit Size:
100mL
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134450 500 mL 500mL
18030350 5 mL 5mL
18060240 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134450 Supplier Toronto Research Chemicals Supplier No. TRCB690030500ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 431-03-8
Molecular Formula C4 H6 O2
Molecular Weight (g/mol) 86.09
Synonym 2,3-Butadione, 2,3-Diketobutane, 2,3-Dioxobutane, Biacetyl, Butanedione, Diacetyl, Dimethyl diketone, Dimethylglyoxal, NSC 8750
IUPAC Name butane-2,3-dione
SMILES CC(=O)C(=O)C

Specifications

Quantity 500 mL
InChI Formula InChI=1S/C4H6O2/c1-3(5)4(2)6/h1-2H3
Formula Weight 86.0368
Chemical Name or Material 2,3-Butanedione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.