missing translation for 'onlineSavingsMsg'
Learn More

2,3-Butanedione, 99%

Product Code. 11492307
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492307 100 g 100g
11484110 10 g 10g
11402317 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492307 Supplier Thermo Scientific Alfa Aesar Supplier No. A14217.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 431-03-8 | C4H6O2 | 86.09 g/mol

2,3-Butanedione is used to form triazine and pteridine ring systems by cyclocondensation with amines. It is also used to inactivate aminopeptidase-N and acts as a precursor to alpha-diones. It finds use in alcoholic beverages, foods, to modify arginyl residues in proteins and to give a buttery flavor to microwave popcorns.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Butanedione is used to form triazine and pteridine ring systems by cyclocondensation with amines. It is also used to inactivate aminopeptidase-N and acts as a precursor to alpha-diones. It finds use in alcoholic beverages, foods, to modify arginyl residues in proteins and to give a buttery flavor to microwave popcorns.

Solubility
Miscible with water.

Notes
Store in a cool place. Incompatible with acids, strong bases, metals, reducing agents and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 431-03-8
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00008756
InChI Key QSJXEFYPDANLFS-UHFFFAOYSA-N
Synonym 2,3-butanedione, diacetyl, biacetyl, dimethylglyoxal, 2,3-diketobutane, dimethyl glyoxal, butanedione, dimethyl diketone, 2,3-butadione, 2,3-dioxobutane
PubChem CID 650
ChEBI CHEBI:16583
IUPAC Name butane-2,3-dione
SMILES CC(=O)C(=O)C

Specifications

Melting Point -3°C
Density 0.983
Boiling Point 87°C to 88°C
Flash Point 10°C (50°F)
Odor Sweet
Refractive Index 1.394
Quantity 100 g
UN Number UN2346
Beilstein 605398
Merck Index 14,2966
Solubility Information Miscible with water.
Formula Weight 86.09
Percent Purity 99%
Chemical Name or Material 2,3-Butanedione
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,3-Butanedione
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 2,3-Butanedione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.