missing translation for 'onlineSavingsMsg'
Learn More

2,3-Butanedione, 99%

Product Code. 10755171 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10755171 500 mL Glass bottle 500mL
10579990 5 mL Glass bottle 5mL
10745171 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10755171 Supplier Thermo Scientific Acros Supplier No. 107655000
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 431-03-8 | C4H6O2 | 86.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 431-03-8
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00008756
InChI Key QSJXEFYPDANLFS-UHFFFAOYSA-N
Synonym 2,3-butanedione, diacetyl, biacetyl, dimethylglyoxal, 2,3-diketobutane, dimethyl glyoxal, butanedione, dimethyl diketone, 2,3-butadione, 2,3-dioxobutane
PubChem CID 650
ChEBI CHEBI:16583
IUPAC Name butane-2,3-dione
SMILES CC(=O)C(=O)C

Especificaciones

CAS Min % 98.5
CAS Max % 100.0
Melting Point -4°C to 2°C
Color Yellow
Density 0.9850g/mL
Boiling Point 88.0°C
Flash Point 7°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3COCOCH3
Refractive Index 1.3910 to 1.3990
Quantity 500 mL
Beilstein 01, 769
Fieser 07,21
Merck Index 15, 2969
Specific Gravity 0.985
Solubility Information Solubility in water: 200g/L (20°C). Other solubilities: miscible with alcohol, ether, very soluble in acetone, soluble in oils, fats, propylene glycol, glycerol, soluble in carbitols, all important organic solven, ts
Formula Weight 86.09
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2,3-Butanedione, 99%
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons compliance-icons
Nombre del producto
  • 2,3-Butanedione
Palabra de advertencia
  • Danger
Clasificación
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Indicaciones de peligro
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H331-Toxic if inhaled.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.