missing translation for 'onlineSavingsMsg'
Learn More

2,3-Butanediol, 98%, mixture of racemic and meso forms, techn.

Product Code. 10428440 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10428440 1 L Glass bottle 1 L
10284510 250 mL Glass bottle 250mL
11944631 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10428440 Supplier Thermo Scientific Acros Supplier No. 107640010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 513-85-9 | C4H10O2 | 90.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-85-9
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
InChI Key OWBTYPJTUOEWEK-UHFFFAOYSA-N
Synonym 2,3-butanediol, 2,3-butylene glycol, 2,3-dihydroxybutane, dimethylethylene glycol, dimethylene glycol, pseudobutylene glycol, sym-dimethylethylene glycol, ccris 5501, dl-2,3-butanediol, 2,3-butanediol, r*,r*-.+/-.
PubChem CID 262
ChEBI CHEBI:62064
IUPAC Name butane-2,3-diol
SMILES CC(C(C)O)O

Specifications

Melting Point 19.0°C
Color Colorless to Yellow
Density 1.0100g/mL
Boiling Point 180.0°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OH)CH(OH)CH3
Refractive Index 1.4330 to 1.4370
Quantity 1 L
Beilstein 01,479
Merck Index 15,1571
Specific Gravity 1.01
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in alcohol, ether and diisopropyl ether
Formula Weight 90.12
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material 2, 3-Butanediol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.