missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Bromophenyl)furan, ≥97%, Thermo Scientific™

Product Code. 10061334
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10061334 1 g Amber Glass Bottle 1g
10634005 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10061334 Supplier Thermo Scientific Maybridge Supplier No. CC46110DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 85553-51-1
Molecular Formula C10H7BrO
Molecular Weight (g/mol) 223.07
MDL Number MFCD07775615
InChI Key YQTIZEZABLIGPW-UHFFFAOYSA-N
Synonym 2-3-bromophenyl furan, 2-3-bromo-phenyl-furan, furan,2-3-bromophenyl
PubChem CID 10609118
IUPAC Name 2-(3-bromophenyl)furan
SMILES BrC1=CC=CC(=C1)C1=CC=CO1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 223.07
Percent Purity 97%
Chemical Name or Material 2-(3-Bromophenyl)furan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.