missing translation for 'onlineSavingsMsg'
Learn More

2-[3-(Bromomethyl)phenyl]thiophene, ≥97%, Thermo Scientific™

Product Code. 10528823
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 85553-44-2
Molecular Formula C11H9BrS
Molecular Weight (g/mol) 253.157
MDL Number MFCD08435867
InChI Key JQQDEBDYUAKNGD-UHFFFAOYSA-N
Synonym 2-3-bromomethyl phenyl thiophene, 3-2-thienyl benzyl bromide, 2-3-bromomethyl phenyl-thiophene
PubChem CID 13150400
IUPAC Name 2-[3-(bromomethyl)phenyl]thiophene
SMILES C1=CC(=CC(=C1)CBr)C2=CC=CS2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 253.16
Percent Purity 97%
Chemical Name or Material 2-[3-(Bromomethyl)phenyl]thiophene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'