missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Bromo-4-fluorophenyl)acetonitrile, 96%

Product Code. 15448808
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448808 1 g 1g
15468808 250 mg 250mg
15458808 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15448808 Supplier Thermo Scientific Alfa Aesar Supplier No. H62260.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 501420-63-9 | C8H5BrFN | 214.037 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 501420-63-9
Molecular Formula C8H5BrFN
Molecular Weight (g/mol) 214.037
MDL Number MFCD08458154
InChI Key ORKCKZRBHXMWBO-UHFFFAOYSA-N
Synonym 2-3-bromo-4-fluorophenyl acetonitrile, 3-bromo-4-fluorophenylacetonitrile, 3-bromo-4-fluorobenzyl cyanide, benzeneacetonitrile, 3-bromo-4-fluoro, 3-bromo-4-fluorophenyl acetonitrile, 3-bromo-4-fluoro benzyl cyanide, 3-bromo-4-fluorobenzeneacetonitrile, 2-3-bromo-4-fluorophenyl ethanenitrile
PubChem CID 20113897
IUPAC Name 2-(3-bromo-4-fluorophenyl)acetonitrile
SMILES C1=CC(=C(C=C1CC#N)Br)F

Specifications

Quantity 1 g
UN Number UN3439
Formula Weight 214.03
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-(3-Bromo-4-fluorophenyl)acetonitrile
compliance-icons
Product Identifier
  • 2-(3-Bromo-4-fluorophenyl)acetonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 2-(3-Bromo-4-fluorophenyl)acetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.