missing translation for 'onlineSavingsMsg'
Learn More

2,3-Bis[(acetyloxy)methyl]-1-oxoquinoxalin-1-ium-4(1H)-olate, TRC

Product Code. 30004118
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004118 50 mg 50mg
30010393 5 mg 5mg
30010416 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004118 Supplier Toronto Research Chemicals Supplier No. TRCB32562050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10103-89-6
Molecular Formula C14 H14 N2 O6
Molecular Weight (g/mol) 306.27
Synonym 2,3-Di(acetoxymethyl)quinoxaline di-N-Oxide;
IUPAC Name [3-(acetoxymethyl)-1,4-dioxo-quinoxaline-1,4-diium-2-yl]methyl acetate
SMILES CC(=O)OCC1=C(COC(=O)C)[N+](=O)c2ccccc2[N+]1=O

Specifications

Analyte or Component Names 2,3-Bis[(acetyloxy)methyl]-1-oxoquinoxalin-1-ium-4(1H)-olate
Shipping Condition Room Temperature
InChI Formula InChI=1S/C14H14N2O6/c1-9(17)21-7-13-14(8-22-10(2)18)16(20)12-6-4-3-5-11(12)15(13)19/h3-6H,7-8H2,1-2H3/q+2
Solution Type Neat
Industry Type Forensics
Quantity 50 mg
Formula Weight 306.0841
Chemical Name or Material 2,3-Bis[(acetyloxy)methyl]-1-oxoquinoxalin-1-ium-4(1H)-olate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.