missing translation for 'onlineSavingsMsg'
Learn More

2,3-Benzofuran, 99%

Product Code. 10371374 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10371374 50 mL Glass bottle 50mL
10519800 10 mL Glass bottle 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10371374 Supplier Thermo Scientific Acros Supplier No. 105480500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 271-89-6 | C8H6O | 118.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 271-89-6
Molecular Formula C8H6O
Molecular Weight (g/mol) 118.14
MDL Number MFCD00005847
InChI Key IANQTJSKSUMEQM-UHFFFAOYSA-N
Synonym benzofuran, 2,3-benzofuran, coumarone, benzo b furan, cumarone, benzofurfuran, 1-oxindene, coumaron, 1-oxidene
PubChem CID 9223
ChEBI CHEBI:35260
IUPAC Name 1-benzofuran
SMILES O1C=CC2=CC=CC=C12

Specifications

CAS Min % 99.4
CAS Max % 100.0
Color Brown-Yellow to Yellow
Density 1.0700g/mL
Boiling Point 173°C to 175°C
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 99.4 % min. (GC)
Packaging Glass bottle
Refractive Index 1.5655 to 1.5675
Quantity 50 mL
Beilstein 17, 54
Merck Index 15, 1090
Specific Gravity 1.07
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in aq. alkaline solutions
Formula Weight 118.13
Percent Purity 99.50%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2, 3-Benzofuran
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,3-Benzofuran
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H351-Suspected of causing cancer.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.